Anti-SELPLG Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 688101]

Supplier: R & D Systems
FAB9961G
102884-102EA 501.82 USD
102884-102
Anti-SELPLG Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 688101]
Antibodies
Type: Primary
Antigen: SELPLG
Clonality: Monoclonal
Clone: 688101
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR