Anti-ABCG2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 5D3]

Supplier: R & D Systems
FAB995G
102884-096EA 543.75 USD
102884-096
Anti-ABCG2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 5D3]
Antibodies
Type: Primary
Antigen: ABCG2
Clonality: Monoclonal
Clone: 5D3
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR