Anti-IL21R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 152512]

Supplier: R & D Systems
FAB9911P-100 FAB9911P-025
102884-092EA 253.08 USD
102884-092 102884-094
Anti-IL21R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 152512]
Antibodies
Type: Primary
Antigen: IL21R
Clonality: Monoclonal
Clone: 152512
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR