Anti-SIGIRR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162201]

Supplier: R & D Systems
FAB990A
220018-041EA 639.2 USD
220018-041
Anti-SIGIRR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162201]
Antibodies
Type: Primary
Antigen: SIGIRR
Clonality: Monoclonal
Clone: 162201
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR