Anti-IL3RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 151231]

Supplier: R & D Systems
FAB983P
220018-040EA 420.83 USD
220018-040
Anti-IL3RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 151231]
Antibodies
Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: 151231
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR