Anti-EPCAM Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 158206]

Supplier: R & D Systems
FAB9601N
102883-842EA 543.75 USD
102883-842
Anti-EPCAM Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 158206]
Antibodies
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: 158206
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR