Anti-EPCAM Mouse Monoclonal Antibody (Fluorescein) [clone: 158206]

Supplier: R & D Systems
FAB9601F
220018-036EA 420.83 USD
220018-036
Anti-EPCAM Mouse Monoclonal Antibody (Fluorescein) [clone: 158206]
Antibodies
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: 158206
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR