Anti-EPCAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 158206]

Supplier: R & D Systems
FAB9601A
220018-035EA 577.02 USD
220018-035
Anti-EPCAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 158206]
Antibodies
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: 158206
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR