Anti-DPP4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 155202]

Supplier: R & D Systems
FAB9541P
220018-034EA 446.86 USD
220018-034
Anti-DPP4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 155202]
Antibodies
Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 155202
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR