Anti-ADAM10 Rat Monoclonal Antibody (Fluorescein) [clone: 139712]

Supplier: R & D Systems
FAB946F
220018-030EA 420.83 USD
220018-030
Anti-ADAM10 Rat Monoclonal Antibody (Fluorescein) [clone: 139712]
Antibodies
Type: Primary
Antigen: ADAM10
Clonality: Monoclonal
Clone: 139712
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR