Anti-TNFRSF25 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 59204]

Supplier: R & D Systems
FAB943P
102883-832EA 543.75 USD
102883-832
Anti-TNFRSF25 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 59204]
Antibodies
Type: Primary
Antigen: TNFRSF25
Clonality: Monoclonal
Clone: 59204
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR