Anti-TNFRSF9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 158332]

Supplier: R & D Systems
FAB937P
102883-828EA 349.97 USD
102883-828
Anti-TNFRSF9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 158332]
Antibodies
Type: Primary
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: 158332
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR