Anti-ADAM17 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 111633]

Supplier: R & D Systems
FAB9301P
220018-023EA 549.54 USD
220018-023
Anti-ADAM17 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 111633]
Antibodies
Type: Primary
Antigen: ADAM17
Clonality: Monoclonal
Clone: 111633
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR