Anti-MERTK Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 125518]

Supplier: R & D Systems
FAB8912P
220018-020EA 594.37 USD
220018-020
Anti-MERTK Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 125518]
Antibodies
Type: Primary
Antigen: MERTK
Clonality: Monoclonal
Clone: 125518
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR