Anti-MERTK Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 125518]

Supplier: R & D Systems
FAB8912N
102884-022EA 577.02 USD
102884-022
Anti-MERTK Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 125518]
Antibodies
Type: Primary
Antigen: MERTK
Clonality: Monoclonal
Clone: 125518
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR