Anti-IL10RB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 90220]

Supplier: R & D Systems
FAB874P
220018-018EA 543.75 USD
220018-018
Anti-IL10RB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 90220]
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Monoclonal
Clone: 90220
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR