Anti-IL10RB Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 90220]

Supplier: R & D Systems
FAB874G
102884-018EA 543.75 USD
102884-018
Anti-IL10RB Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 90220]
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Monoclonal
Clone: 90220
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR