Anti-IL10RB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 90220]

Supplier: R & D Systems
FAB874A
102884-016EA 577.02 USD
102884-016
Anti-IL10RB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 90220]
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Monoclonal
Clone: 90220
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR