Anti-LEPR Mouse Monoclonal Antibody (Fluorescein) [clone: 52263]

Supplier: R & D Systems
FAB867F
220018-016EA 420.83 USD
220018-016
Anti-LEPR Mouse Monoclonal Antibody (Fluorescein) [clone: 52263]
Antibodies
Type: Primary
Antigen: LEPR
Clonality: Monoclonal
Clone: 52263
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR