Anti-CDH3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104805]

Supplier: R & D Systems
FAB861P
220018-014EA 543.75 USD
220018-014
Anti-CDH3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104805]
Antibodies
Type: Primary
Antigen: CDH3
Clonality: Monoclonal
Clone: 104805
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR