Anti-AXL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 175128]

Supplier: R & D Systems
FAB8541A
102883-994EA 594.37 USD
102883-994
Anti-AXL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 175128]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 175128
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR