Anti-IL18R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 70625]

Supplier: R & D Systems
FAB840P-100 FAB840P-025
102883-992EA 527.85 USD
102883-992 102883-990
Anti-IL18R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 70625]
Antibodies
Type: Primary
Antigen: IL18R1
Clonality: Monoclonal
Clone: 70625
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR