Anti-IL18R1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 70625]

Supplier: R & D Systems
FAB840G-100 FAB840G-025
102883-988EA 561.11 USD
102883-988 102883-986
Anti-IL18R1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 70625]
Antibodies
Type: Primary
Antigen: IL18R1
Clonality: Monoclonal
Clone: 70625
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR