Anti-IL18R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 70625]

Supplier: R & D Systems
FAB840A-025 FAB840A-100
102883-978EA 274.77 USD
102883-978 102883-980
Anti-IL18R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 70625]
Antibodies
Type: Primary
Antigen: IL18R1
Clonality: Monoclonal
Clone: 70625
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR