Anti-IL12RB1 Mouse Monoclonal Antibody (Fluorescein) [clone: 69310]

Supplier: R & D Systems
FAB839F-025 FAB839F-100
102883-968EA 485.91 USD
102883-968 102883-966
Anti-IL12RB1 Mouse Monoclonal Antibody (Fluorescein) [clone: 69310]
Antibodies
Type: Primary
Antigen: IL12RB1
Clonality: Monoclonal
Clone: 69310
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR