Anti-IL12RB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 69310]

Supplier: R & D Systems
FAB839A-100 FAB839A-025
102883-958EA 271.88 USD
102883-958 102883-960
Anti-IL12RB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 69310]
Antibodies
Type: Primary
Antigen: IL12RB1
Clonality: Monoclonal
Clone: 69310
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR