Anti-SLC16A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 882616]

Supplier: R & D Systems
FAB8275P
102883-898EA 561.11 USD
102883-898
Anti-SLC16A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 882616]
Antibodies
Type: Primary
Antigen: SLC16A1
Clonality: Monoclonal
Clone: 882616
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR