Anti-SLC16A1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 882616]

Supplier: R & D Systems
FAB8275A
102883-896EA 594.37 USD
102883-896
Anti-SLC16A1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 882616]
Antibodies
Type: Primary
Antigen: SLC16A1
Clonality: Monoclonal
Clone: 882616
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR