Anti-MUC4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 781631]

Supplier: R & D Systems
FAB8195A
102883-874EA 594.37 USD
102883-874
Anti-MUC4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 781631]
Antibodies
Type: Primary
Antigen: MUC4
Clonality: Monoclonal
Clone: 781631
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR