Anti-SEMA4A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 757129]

Supplier: R & D Systems
FAB8166A
102883-866EA 552.43 USD
102883-866
Anti-SEMA4A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 757129]
Antibodies
Type: Primary
Antigen: SEMA4A
Clonality: Monoclonal
Clone: 757129
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR