Anti-CD4 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB8165G
102883-864EA 342.74 USD
102883-864
Anti-CD4 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR