Anti-FLT3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 66903]

Supplier: R & D Systems
FAB812A
102883-850EA 561.11 USD
102883-850
Anti-FLT3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 66903]
Antibodies
Type: Primary
Antigen Symbol: FLT3
Antigen Name: fms-related tyrosine kinase 3
Antigen Synonyms: CD135, FLK-2, FLK2, STK1
Clonality: Monoclonal
Clone: 66903
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR