Anti-CXCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 866614]

Supplier: R & D Systems
FAB8110P
102883-504EA 485.91 USD
102883-504
Anti-CXCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 866614]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 866614
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR