Anti-CXCR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 868013]

Supplier: R & D Systems
FAB8109P
102883-500EA 561.11 USD
102883-500
Anti-CXCR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 868013]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 868013
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR