Anti-CXCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 868013]

Supplier: R & D Systems
FAB8109A
102883-496EA 608.83 USD
102883-496
Anti-CXCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 868013]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 868013
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR