Anti-PLAUR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 62022]

Supplier: R & D Systems
FAB807P
220018-001EA 527.85 USD
220018-001
Anti-PLAUR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 62022]
Antibodies
Type: Primary
Antigen: PLAUR
Clonality: Monoclonal
Clone: 62022
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR