Anti-LGR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 707042]

Supplier: R & D Systems
FAB8078A
102883-848EA 594.37 USD
102883-848
Anti-LGR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 707042]
Antibodies
Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 707042
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR