Anti-TM4SF4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 832441]

Supplier: R & D Systems
FAB7998A
102883-820EA 594.37 USD
102883-820
Anti-TM4SF4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 832441]
Antibodies
Type: Primary
Antigen: TM4SF4
Clonality: Monoclonal
Clone: 832441
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR