Anti-SIGLEC8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 837535]

Supplier: R & D Systems
FAB7975P
102883-818EA 566.89 USD
102883-818
Anti-SIGLEC8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 837535]
Antibodies
Type: Primary
Antigen: SIGLEC8
Clonality: Monoclonal
Clone: 837535
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR