Anti-SIGLEC8 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 837535]

Supplier: R & D Systems
FAB7975C
102883-812EA 633.42 USD
102883-812
Anti-SIGLEC8 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 837535]
Antibodies
Type: Primary
Antigen: SIGLEC8
Clonality: Monoclonal
Clone: 837535
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR