Anti-TIGIT Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 741182]

Supplier: R & D Systems
FAB7898N
102883-798EA 594.37 USD
102883-798
Anti-TIGIT Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 741182]
Antibodies
Type: Primary
Antigen: TIGIT
Clonality: Monoclonal
Clone: 741182
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR