Anti-TIGIT Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 741182]

Supplier: R & D Systems
FAB7898A
102883-796EA 639.2 USD
102883-796
Anti-TIGIT Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 741182]
Antibodies
Type: Primary
Antigen: TIGIT
Clonality: Monoclonal
Clone: 741182
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR