Anti-DCSTAMP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 788524]

Supplier: R & D Systems
FAB7824A
102883-786EA 577.02 USD
102883-786
Anti-DCSTAMP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 788524]
Antibodies
Type: Primary
Antigen: DCSTAMP
Clonality: Monoclonal
Clone: 788524
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR