Anti-MS4A4A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 818112]

Supplier: R & D Systems
FAB7797P
102883-564EA 561.11 USD
102883-564
Anti-MS4A4A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 818112]
Antibodies
Type: Primary
Antigen: MS4A4A
Clonality: Monoclonal
Clone: 818112
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR