Anti-ACVR1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8156]

Supplier: R & D Systems
FAB77491A
102883-560EA 566.89 USD
102883-560
Anti-ACVR1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8156]
Antibodies
Type: Primary
Antigen: ACVR1C
Clonality: Monoclonal
Clone: 8156
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR