Anti-PD1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 766104]

Supplier: R & D Systems
FAB7738P
102883-782EA 513.38 USD
102883-782
Anti-PD1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 766104]
Antibodies
Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: 766104
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR