Anti-IL1R1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 129304]

Supplier: R & D Systems
FAB7712P-100 FAB7712P-025
102883-768EA 253.08 USD
102883-768 102883-770
Anti-IL1R1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 129304]
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 129304
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR