Anti-FGFR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 136334]

Supplier: R & D Systems
FAB766A
102883-760EA 633.42 USD
102883-760
Anti-FGFR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 136334]
Antibodies
Type: Primary
Antigen Symbol: FGFR3
Antigen Name: fibroblast growth factor receptor 3
Antigen Synonyms: ACH, CD333, CEK2, HSFGFR3EX, JTK4
Clonality: Monoclonal
Clone: 136334
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR