Anti-CDH3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 106020]

Supplier: R & D Systems
FAB761P
220017-987EA 513.38 USD
220017-987
Anti-CDH3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 106020]
Antibodies
Type: Primary
Antigen: CDH3
Clonality: Monoclonal
Clone: 106020
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR