Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 745201]

Supplier: R & D Systems
FAB7616P
102883-752EA 485.91 USD
102883-752
Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 745201]
Antibodies
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: 745201
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR